Imprimer

ALFAB25721

B25721.30EA 82.7 CHF
B25721.30
ALFAB25721
Strongly acidic cation-exchange resin, 550-650 micron beads; minimum exchange capacity 1.8 meq/mL.
Applications: It is designed for demineralization using HCl or H2SO4 as regenerate in co-current or counter-current operation.
Store in cool, dry place in a well sealed container. Store away from oxidizing agents

Solubility: Insoluble in water.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR