Imprimer

Iron tris(trifluoromethylsulphonyl)imide

Danger

47186.MDEA 77 CHF
47186.MD
Iron tris(trifluoromethylsulphonyl)imide
Iron tris(trifluoromethylsulphonyl)imide
Formule: C₆F₁₈FeN₃O₁₂S₆
MW: 896,25 g/mol
température de stockage: Température ambiante
MDL Number: MFCD23380171
Numéro CAS: 207861-59-4
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR