Aluminum manganese (alloy 3003), tube 7.14mm (0.281in) OD, 6.43mm (0.253in) ID
45942.RMEA
54.5
CHF
45942.RM
Aluminum manganese (alloy 3003), tube 7.14mm (0.281in) OD, 6.43mm (0.253in) ID
Aluminum manganese (alloy 3003)
Stronger aluminum manganese alloy, N3 is specified for troughed and corrugated sheets.
Anodized aluminum manganese alloy is widely used for curtain walling