FAM benzine DIN 51635, Supelco®
|
Danger
|
M-Clarity™ (Niveau de qualité) = MQ200
1.01771.1000EA
117
CHF
1.01771.1000
FAM benzine DIN 51635, Supelco®
FAM benzine
Boiling point: 65 °C (1013 hPa)
Density: 0.76 g/cm3 (15 °C)
Explosion limit: 0.7 - 8.3 %(V)
Flash point: -26 °C
Ignition temperature: 245 °C
Melting Point: <-30 °C
Vapor pressure: 10 hPa (20 °C)
Solubility: NULL g/l slightly soluble
température de stockage:
Température ambiante |
MDL Number:
MFCD00282001 Numéro CAS: 64742-49-0 ONU: 1268 ADR: 3,II |
Résultats des tests de spécification
Colour | ≤ 10 Hazen |
Density (15°C/DIN 51757) | 0.690 - 0.710 |
Boiling range (DIN 51751 (60-90°C)) | 85 - 95 % (v/v) |
Aniline point (DIN ISO 2977) | 59.0 - 61.0 °C |
Aromatics (as benzene) | ≤ 0.1 % |
n-Hexane (GC) | ≤ 5.0 % |
Non volatile matter | ≤ 0.002 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...