Imprimer

Anti-ABHD17C Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fournisseur: US Biological
USBI035357-FITCEA 1200 CHF
USBI035357-FITC
Anti-ABHD17C Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: ABHD17C (abhydrolase domain containing 17C)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR