Imprimer

Anti-FAIM2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fournisseur: US Biological
USBI035347-APCEA 1200 CHF
USBI035347-APC
Anti-FAIM2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: FAIM2 (Fas apoptotic inhibitory molecule 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR