Imprimer

Anti-EZH2 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: United States Biological
USBI035290-BIOTINEA 1200 CHF
USBI035290-BIOTIN
Anti-EZH2 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: EZH2 (enhancer of zeste homolog 2 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR