Imprimer

Anti-EZH2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fournisseur: United States Biological
USBI035287-PEEA 1200 CHF
USBI035287-PE
Anti-EZH2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50
Do not freeze!

Type: Primary
Antigen: EZH2 (enhancer of zeste homolog 2 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR