Imprimer

Anti-EYA1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fournisseur: United States Biological
USBI035283-APCEA 1200 CHF
USBI035283-APC
Anti-EYA1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: EYA1 (eyes absent homolog 1 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR