Imprimer

Anti-EXD3 Rabbit Polyclonal Antibody

Fournisseur: United States Biological
USBI035264EA 825 CHF
USBI035264
Anti-EXD3 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: EXD3 (exonuclease 3'-5' domain containing 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR