Imprimer

Anti-ETV6 Rabbit Polyclonal Antibody

Fournisseur: United States Biological
USBI035254EA 825 CHF
USBI035254
Anti-ETV6 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ETV6 (ets variant 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR