Imprimer

Anti-PRSS48 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: United States Biological
USBI035243-BIOTINEA 1200 CHF
USBI035243-BIOTIN
Anti-PRSS48 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: PRSS48 (protease, serine, 48)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR