Imprimer

Anti-ESR2 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: United States Biological
USBI035240-BIOTINEA 1200 CHF
USBI035240-BIOTIN
Anti-ESR2 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ESR2 (estrogen receptor 2 (ER beta))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR