Imprimer

Anti-ESR1 isoform4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fournisseur: United States Biological
USBI035235-PEEA 1200 CHF
USBI035235-PE
Anti-ESR1 isoform4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ESR1 isoform4
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR