Imprimer

Anti-ESR1 isoform1 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: United States Biological
USBI035234-BIOTINEA 1200 CHF
USBI035234-BIOTIN
Anti-ESR1 isoform1 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ESR1 isoform1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR