Imprimer

Anti-ERI2 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: US Biological
USBI035207-BIOTINEA 1200 CHF
USBI035207-BIOTIN
Anti-ERI2 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ERI2 (ERI1 exoribonuclease family member 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR