Imprimer

Anti-ERBB2 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: US Biological
USBI035187-BIOTINEA 1200 CHF
USBI035187-BIOTIN
Anti-ERBB2 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Dot Blot 1:500

Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR