Imprimer

Anti-EPS8L1 Rabbit Polyclonal Antibody

Fournisseur: US Biological
USBI035133EA 825 CHF
USBI035133
Anti-EPS8L1 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: EPS8L1 (EPS8-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR