Imprimer

Anti-EPOR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fournisseur: US Biological
USBI035132-FITCEA 1200 CHF
USBI035132-FITC
Anti-EPOR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Type: Primary
Antigen: EPOR (erythropoietin receptor)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR