Imprimer

Anti-EPHB4 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fournisseur: US Biological
USBI035127-APEA 1200 CHF
USBI035127-AP
Anti-EPHB4 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: EPHB4 (EPH receptor B4)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR