Imprimer

Anti-EPHA7 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fournisseur: US Biological
USBI035122-APCEA 1200 CHF
USBI035122-APC
Anti-EPHA7 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorps
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: EPHA7 (EPH receptor A7)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR