Imprimer

Anti-EPHA7 Rabbit Polyclonal Antibody

Fournisseur: US Biological
USBI035122EA 825 CHF
USBI035122
Anti-EPHA7 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: EPHA7 (EPH receptor A7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR