Imprimer

Anti-EPCAM Rabbit Polyclonal Antibody

Fournisseur: US Biological
USBI035110EA 825 CHF
USBI035110
Anti-EPCAM Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: Western Blot: 1:1000; Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR