Imprimer

Anti-EPB41L5 Rabbit Polyclonal Antibody

Fournisseur: US Biological
USBI035106EA 825 CHF
USBI035106
Anti-EPB41L5 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: EPB41L5 (erythrocyte membrane protein band 4.1 like 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR