Imprimer

Anti-EPAS1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fournisseur: US Biological
USBI035102-FITCEA 1200 CHF
USBI035102-FITC
Anti-EPAS1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: EPAS1 (endothelial PAS domain protein 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR