Imprimer

Anti-ENPP6 Rabbit Polyclonal Antibody

Fournisseur: US Biological
USBI035085EA 825 CHF
USBI035085
Anti-ENPP6 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: ENPP6 (ectonucleotide pyrophosphatase/phosphodiesterase 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR