Imprimer

Anti-ENPP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: US Biological
USBI035083-HRPEA 1200 CHF
USBI035083-HRP
Anti-ENPP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ENPP4 (ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR