Imprimer

Anti-EIF3CL Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fournisseur: US Biological
USBI035003-APEA 1200 CHF
USBI035003-AP
Anti-EIF3CL Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: EIF3CL (eukaryotic translation initiation factor 3, subunit C-like)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR