Imprimer

Anti-EIF3A Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fournisseur: US Biological
USBI035000-PEEA 1200 CHF
USBI035000-PE
Anti-EIF3A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: EIF3A (eukaryotic translation initiation factor 3, subunit A)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR