Imprimer

Anti-EIF2AK3 Rabbit Polyclonal Antibody

Fournisseur: US Biological
USBI034988EA 825 CHF
USBI034988
Anti-EIF2AK3 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: EIF2AK3 (eukaryotic translation initiation factor 2-alpha kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR