Imprimer

Anti-EIF2A Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fournisseur: US Biological
USBI034985-PEEA 1200 CHF
USBI034985-PE
Anti-EIF2A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: EIF2A (eukaryotic translation initiation factor 2A, 65kDa)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR