Imprimer

Anti-EGFR Rabbit Polyclonal Antibody

Fournisseur: US Biological
USBI034960EA 825 CHF
USBI034960
Anti-EGFR Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:50-100

Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR