Imprimer

Anti-EGF Rabbit Polyclonal Antibody (Biotin)

Fournisseur: US Biological
USBI034958-BIOTINEA 1200 CHF
USBI034958-BIOTIN
Anti-EGF Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: EGF (epidermal growth factor)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR