Imprimer

Anti-EFEMP2 Rabbit Polyclonal Antibody

Fournisseur: US Biological
USBI034941EA 825 CHF
USBI034941
Anti-EFEMP2 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: EFEMP2 (EGF containing fibulin-like extracellular matrix protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR