Imprimer

Anti-EEF2K Rabbit Polyclonal Antibody

Fournisseur: US Biological
USBI034930EA 825 CHF
USBI034930
Anti-EEF2K Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: EEF2K (eukaryotic elongation factor-2 kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR