Imprimer

Anti-EEF1G Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fournisseur: US Biological
USBI034928-APCEA 1200 CHF
USBI034928-APC
Anti-EEF1G Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: EEF1G (eukaryotic translation elongation factor 1 gamma)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR