Imprimer

Anti-EDN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fournisseur: US Biological
USBI034920-APCEA 1200 CHF
USBI034920-APC
Anti-EDN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: EDN1 (endothelin 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR