Imprimer

Anti-EDEM3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fournisseur: US Biological
USBI034914-APCEA 1200 CHF
USBI034914-APC
Anti-EDEM3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: EDEM3 (ER degradation enhancer, mannosidase alpha-like 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR