Imprimer

Anti-ECSCR Rabbit Polyclonal Antibody (Biotin)

Fournisseur: US Biological
USBI034907-BIOTINEA 1200 CHF
USBI034907-BIOTIN
Anti-ECSCR Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ECSCR (endothelial cell surface expressed chemotaxis and apoptosis regulator)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR