Imprimer

Anti-S1PR4 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41421-100EA 0 CHF
BIRBORB41421-100
Anti-S1PR4 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000

Type: Primary
Antigen: S1PR4 (sphingosine-1-phosphate receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR