Imprimer

Anti-IPO13 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41389-100EA 0 CHF
BIRBORB41389-100
Anti-IPO13 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: IPO13 (importin 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR