Imprimer

Anti-DYRK3 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41380-100EA 353 CHF
BIRBORB41380-100
Anti-DYRK3 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500

Type: Primary
Antigen: DYRK3 (dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR