Imprimer
Anti-PIK3R3 Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
BIRBORB41375-100EA 0 CHF
BIRBORB41375-100
Anti-PIK3R3 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000
Type: Primary
Antigen: PIK3R3 (phosphoinositide-3-kinase, regulatory subunit 3 (gamma))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human