Imprimer

Anti-ZMYND10 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41369-100EA 0 CHF
BIRBORB41369-100
Anti-ZMYND10 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000

Type: Primary
Antigen: ZMYND10 (zinc finger, MYND-type containing 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR