Imprimer

Anti-GLE1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41317-100EA 0 CHF
BIRBORB41317-100
Anti-GLE1 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: GLE1 (GLE1 RNA export mediator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR