Imprimer

Anti-CAMK1D Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41138-100EA 424 CHF
BIRBORB41138-100
Anti-CAMK1D Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: CAMK1D (calcium/calmodulin-dependent protein kinase ID)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR