Imprimer

Anti-SOCS4 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41093-100EA 0 CHF
BIRBORB41093-100
Anti-SOCS4 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:100-1:1000

Type: Primary
Antigen: SOCS4 (suppressor of cytokine signaling 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR