Imprimer

Anti-interferon alpha receptor 1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB40949-100EA 353 CHF
BIRBORB40949-100
Anti-interferon alpha receptor 1 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000

Type: Primary
Antigen: interferon alpha receptor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR