Imprimer

Anti-NR3C1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB40818-100EA 0 CHF
BIRBORB40818-100
Anti-NR3C1 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000

Type: Primary
Antigen: NR3C1 (nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR