Imprimer

Anti-LCN2 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27771-100EA 526 CHF
BIRBORB27771-100
Anti-LCN2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR